missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HYPB Monoclonal antibody specifically detects HYPB in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | HYPB |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL9956 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ22472, FLJ23184, FLJ45883, FLJ46217, HIF1, HIF-1EC 2.1.1.43, HIP-1, histone-lysine N-methyltransferase SETD2, hSET2, huntingtin interacting protein 1, Huntingtin yeast partner B, Huntingtin-interacting protein 1, Huntingtin-interacting protein B, HYPBp231HBP, KIAA1732SET domain-containing protein 2, KMT3AHBP231, Lysine N-methyltransferase 3A, SET domain containing 2, SET2FLJ16420 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from Q9BYW2, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?