Description
PHYH Monoclonal specifically detects PHYH in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Gene Alias | LNAP1, PAHXLN1, peroxisomal, PhyH, PHYH1, phytanoyl-CoA 2-hydroxylase, phytanoyl-CoA hydroxylase, phytanoyl-CoA hydroxylase (Refsum disease), RD |
| Content And Storage | Store at 4°C in the dark. |
| Product Type | Antibody |
| Product Line | Novus |