Learn More
HYDRANAL™ - Titrant 2 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™
Product Code. p-7122064 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15624360 1 L 1 L
15634360 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15624360 Supplier Honeywell-Hydranal Supplier No. 347231L
Request Bulk or Custom Format
Specifications
Chemical Name or Material HYDRANAL™-Titrant 2 E
Density 0.83g/mL
Quantity 1 L
Flash Point 12°C (53.6°F)
UN Number UN1170
compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Titrant 2 E
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol; ethyl alcohol, iodine, 1H-Imidazole monohydriodide