missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronidase 4/HYAL4 Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30576593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30576593 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30576593 Supplier Novus Biologicals Supplier No. NBP346062CL1
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hyaluronidase 4/HYAL4 Polyclonal antibody specifically detects Hyaluronidase 4/HYAL4 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hyaluronidase 4/HYAL4
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias chondroitin sulfate hydrolase, CSHY, EC 3.2.1, EC 3.2.1.35, Hyal-4, hyaluronidase 4, hyaluronidase-4, hyaluronoglucosaminidase 4, hyaluronoglucosaminidase-4
Host Species Rabbit
Immunogen hyaluronoglucosaminidase 4. (Uniprot# Q2M3T9)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23553
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.