missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuR/ELAVL1 Antibody (OTI14F4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02287
This item is not returnable.
View return policy
Description
HuR/ELAVL1 Monoclonal specifically detects HuR/ELAVL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HuR/ELAVL1 | |
| Monoclonal | |
| 0.78 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 | |
| ELAVL1 | |
| Full length human recombinant protein of human ELAVL1(NP_001410) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI14F4 | |
| Unconjugated | |
| ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R), ELAV-like protein 1, embryonic lethal, abnormal vision, drosophila, homolog-like 1, Hu antigen R, Hua, Hu-antigen R, HuR, HURELAV1, MelG | |
| Mouse | |
| 36.1 kDa | |
| 0.1 mL | |
| Vision | |
| 1994 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction