missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Humanin Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-245H
This item is not returnable.
View return policy
Description
Humanin Polyclonal antibody specifically detects Humanin in Human samples. It is validated for Western Blot
Specifications
| Humanin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 100462977 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| HN, HUNIN | |
| Residues 9-24 (LLLLTSEIDLPVKRRA) of the human Humanin protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction