Learn More
Abnova™ Human WISP1 Full-length ORF (NP_543028.1, 1 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008840-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like domain. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. It is expressed at a high level in fibroblast cells, and overexpressed in colon tumors. The encoded protein binds to decorin and biglycan, two members of a family of small leucine-rich proteoglycans present in the extracellular matrix of connective tissue, and possibly prevents the inhibitory activity of decorin and biglycan in tumor cell proliferation. It also attenuates p53-mediated apoptosis in response to DNA damage through activation of the Akt kinase. It is 83% identical to the mouse protein at the amino acid level. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq]
Sequence: MRWFLPWTLAAVTAAAASTVLATALSPAPTTMDFTPAPLEDTSSRPQFCKWPCECPPSPPRCPLGVSLITDGCECCKMCAQQLGDNCTEAAICDPHRGLYCDYSGDRPRYAIGVCAHAVGEVEAWHRNCIAYTSPWSPCSTSCGLGVSTRISNVNAQCWPEQESRLCNLRPCDVDIHTLIKAGKKCLAVYQPEASMNFTLAGCISTRSYQPKYCGVCMDNRCCIPYKSKTIDVSFQCPDGLGFSRQVLWINACFCNLSCRNPNDIFADLESYPDFSEIANSpecifications
NP_543028.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCN4/WISP1c/WISP1i/WISP1tc | |
WISP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8840 | |
WISP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRWFLPWTLAAVTAAAASTVLATALSPAPTTMDFTPAPLEDTSSRPQFCKWPCECPPSPPRCPLGVSLITDGCECCKMCAQQLGDNCTEAAICDPHRGLYCDYSGDRPRYAIGVCAHAVGEVEAWHRNCIAYTSPWSPCSTSCGLGVSTRISNVNAQCWPEQESRLCNLRPCDVDIHTLIKAGKKCLAVYQPEASMNFTLAGCISTRSYQPKYCGVCMDNRCCIPYKSKTIDVSFQCPDGLGFSRQVLWINACFCNLSCRNPNDIFADLESYPDFSEIAN | |
RUO | |
WISP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |