Learn More
Abnova™ Human WISP1 Full-length ORF (NP_543028.1, 1 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_543028.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8840 |
Molecular Weight (g/mol) | 57.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102972
|
Abnova™
H00008840-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16112972
|
Abnova™
H00008840-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like domain. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. It is expressed at a high level in fibroblast cells, and overexpressed in colon tumors. The encoded protein binds to decorin and biglycan, two members of a family of small leucine-rich proteoglycans present in the extracellular matrix of connective tissue, and possibly prevents the inhibitory activity of decorin and biglycan in tumor cell proliferation. It also attenuates p53-mediated apoptosis in response to DNA damage through activation of the Akt kinase. It is 83% identical to the mouse protein at the amino acid level. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq]
Sequence: MRWFLPWTLAAVTAAAASTVLATALSPAPTTMDFTPAPLEDTSSRPQFCKWPCECPPSPPRCPLGVSLITDGCECCKMCAQQLGDNCTEAAICDPHRGLYCDYSGDRPRYAIGVCAHAVGEVEAWHRNCIAYTSPWSPCSTSCGLGVSTRISNVNAQCWPEQESRLCNLRPCDVDIHTLIKAGKKCLAVYQPEASMNFTLAGCISTRSYQPKYCGVCMDNRCCIPYKSKTIDVSFQCPDGLGFSRQVLWINACFCNLSCRNPNDIFADLESYPDFSEIANSpecifications
NP_543028.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MRWFLPWTLAAVTAAAASTVLATALSPAPTTMDFTPAPLEDTSSRPQFCKWPCECPPSPPRCPLGVSLITDGCECCKMCAQQLGDNCTEAAICDPHRGLYCDYSGDRPRYAIGVCAHAVGEVEAWHRNCIAYTSPWSPCSTSCGLGVSTRISNVNAQCWPEQESRLCNLRPCDVDIHTLIKAGKKCLAVYQPEASMNFTLAGCISTRSYQPKYCGVCMDNRCCIPYKSKTIDVSFQCPDGLGFSRQVLWINACFCNLSCRNPNDIFADLESYPDFSEIAN | |
RUO | |
WISP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8840 | |
WISP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCN4/WISP1c/WISP1i/WISP1tc | |
WISP1 | |
Recombinant | |
wheat germ expression system |