Learn More
Abnova™ Human TSN Full-length ORF (NP_004613.1, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004613.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7247 |
Molecular Weight (g/mol) | 52.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16103838
|
Abnova™
H00007247-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16113838
|
Abnova™
H00007247-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a DNA-binding protein which specifically recognizes conserved target sequences at the breakpoint junction of chromosomal translocations. Translin polypeptides form a multimeric structure that is responsible for its DNA-binding activity. Recombination-associated motifs and translin-binding sites are present at recombination hotspots and may serve as indicators of breakpoints in genes which are fused by translocations. These binding activities may play a crucial role in chromosomal translocation in lymphoid neoplasms. [provided by RefSeq]
Sequence: MSVSEIFVELQGFLAAEQDIREEIRKVVQSLEQTAREILTLLQGVHQGAGFQDIPKRCLKAREHFGTVKTHLTSLKTKFPAEQYYRFHEHWRFVLQRLVFLAAFVVYLETETLVTREAVTEILGIEPDREKGFHLDVEDYLSGVLILASELSRLSVNSVTAGDYSRPLHISTFINELDSGFRLLNLKNDSLRKRYDGLKYDVKKVEEVVYDLSIRGFNKETAAACVEKSpecifications
NP_004613.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSVSEIFVELQGFLAAEQDIREEIRKVVQSLEQTAREILTLLQGVHQGAGFQDIPKRCLKAREHFGTVKTHLTSLKTKFPAEQYYRFHEHWRFVLQRLVFLAAFVVYLETETLVTREAVTEILGIEPDREKGFHLDVEDYLSGVLILASELSRLSVNSVTAGDYSRPLHISTFINELDSGFRLLNLKNDSLRKRYDGLKYDVKKVEEVVYDLSIRGFNKETAAACVEK | |
RUO | |
TSN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7247 | |
TSN (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCLF-1/REHF-1/TRSLN | |
TSN | |
Recombinant | |
wheat germ expression system |