Learn More
Abnova™ Human TSN Full-length ORF (NP_004613.1, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007247-P02.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a DNA-binding protein which specifically recognizes conserved target sequences at the breakpoint junction of chromosomal translocations. Translin polypeptides form a multimeric structure that is responsible for its DNA-binding activity. Recombination-associated motifs and translin-binding sites are present at recombination hotspots and may serve as indicators of breakpoints in genes which are fused by translocations. These binding activities may play a crucial role in chromosomal translocation in lymphoid neoplasms. [provided by RefSeq]
Sequence: MSVSEIFVELQGFLAAEQDIREEIRKVVQSLEQTAREILTLLQGVHQGAGFQDIPKRCLKAREHFGTVKTHLTSLKTKFPAEQYYRFHEHWRFVLQRLVFLAAFVVYLETETLVTREAVTEILGIEPDREKGFHLDVEDYLSGVLILASELSRLSVNSVTAGDYSRPLHISTFINELDSGFRLLNLKNDSLRKRYDGLKYDVKKVEEVVYDLSIRGFNKETAAACVEKSpecifications
NP_004613.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCLF-1/REHF-1/TRSLN | |
TSN | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7247 | |
TSN (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVSEIFVELQGFLAAEQDIREEIRKVVQSLEQTAREILTLLQGVHQGAGFQDIPKRCLKAREHFGTVKTHLTSLKTKFPAEQYYRFHEHWRFVLQRLVFLAAFVVYLETETLVTREAVTEILGIEPDREKGFHLDVEDYLSGVLILASELSRLSVNSVTAGDYSRPLHISTFINELDSGFRLLNLKNDSLRKRYDGLKYDVKKVEEVVYDLSIRGFNKETAAACVEK | |
RUO | |
TSN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |