missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRIM38 Antibody, R&D Systems™

Description
TRIM38 Monoclonal specifically detects TRIM38 in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
| Antigen | TRIM38 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 933011 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | O00635 |
| Gene Alias | MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, RoRet, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet |
| Gene Symbols | TRIM38 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?