missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TRDMT1 Full-length ORF (AAH47733.1, 1 a.a. - 391 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH47733.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1787 |
Molecular Weight (g/mol) | 71kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16164131
|
Abnova™
H00001787-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16154131
|
Abnova™
H00001787-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein responsible for the methylation of aspartic acid transfer RNA. While similar in sequence and structure to DNA cytosine methyltransferases, this gene is distinct and highly conserved in its function among taxa. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFPHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVENKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYESpecifications
AAH47733.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFPHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVENKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYE | |
RUO | |
TRDMT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1787 | |
TRDMT1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DNMT2/M.HsaIIP/PuMet/RNMT1 | |
TRDMT1 | |
Recombinant | |
wheat germ expression system |