Learn More
Abnova™ Human TRDMT1 Full-length ORF (AAH47733.1, 1 a.a. - 391 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001787-P02.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein responsible for the methylation of aspartic acid transfer RNA. While similar in sequence and structure to DNA cytosine methyltransferases, this gene is distinct and highly conserved in its function among taxa. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFPHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVENKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYESpecifications
AAH47733.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DNMT2/M.HsaIIP/PuMet/RNMT1 | |
TRDMT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1787 | |
TRDMT1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFPHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVENKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYE | |
RUO | |
TRDMT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |