missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Trappin-2/Elafin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF1747
This item is not returnable.
View return policy
Description
Trappin-2/Elafin/Skalp Polyclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Trappin-2/Elafin/Skalp | |
| Polyclonal | |
| Unconjugated | |
| P19957 | |
| PI3 | |
| Mouse myeloma cell line NS0-derived recombinant human Trappin-2/Elafin Ala23-Gln117 Accession # P19957 | |
| 100 μg | |
| Primary | |
| Detects human Trappin-2/Elafin in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) SLPI, rhTGM-2, and human Neutrophil Elastase is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Neutralization, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 15.6 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| cementoin, Elafin, Elastase-specific inhibitor, ESI, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI3, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALP, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, Trappin2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5266 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction