missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNPO2 Partial ORF (NP_038461, 201 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00030000-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106246

  • 528.00€ / 25µg

Estimated Shipment: 18-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MDRAQALMDNIDTFIEHLFALAVDDDPEVRKNVCRALVMLLEVRIDRLIPHMHSIIQYMLQRTQDHDENVALEACEFWLTLAEQPICKEVLASHLVQLIP
Specifications

Specifications

NP_038461
Liquid
30000
TNPO2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12155/IPO3/KPNB2B/TRN2
TNPO2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDRAQALMDNIDTFIEHLFALAVDDDPEVRKNVCRALVMLLEVRIDRLIPHMHSIIQYMLQRTQDHDENVALEACEFWLTLAEQPICKEVLASHLVQLIP
RUO
TNPO2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers