missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNNI3K (NP_057062.1, 1 a.a. - 727 a.a.) Partial Recombinant Protein with His tag expressed in baculovirus infected Sf21 cells

Product Code. 16048266
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16048266 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16048266 Supplier Abnova™ Supplier No. P5560.5ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Specifications

Accession Number NP_057062.1
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 51086
Molecular Weight (g/mol) 108kDa
Name TNNI3K (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing Loading 1 ug protein in SDS-PAGE
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CARK/MGC142099/MGC33828
Common Name TNNI3K
Gene Symbol TNNI3K
Biological Activity The activity was determined by ELISA. The enzyme was incubated with biotinylated peptide in streptavidin-coated ELISA plate. Phosphorylation was detected by HRP-labeled anti-phospho antibody. Substrate: Poly-(GT). ATP: 100μM.
Species Baculovirus
Recombinant Recombinant
Protein Tag His
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 97% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.