missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TNK1 Partial ORF (NP_003976, 559 a.a. - 666 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_003976 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8711 |
Molecular Weight (g/mol) | 37.62kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16149085
|
Abnova™
H00008711-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16159085
|
Abnova™
H00008711-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
TNK1 is a nonreceptor tyrosine kinase. These kinases, like members of the SRC (MIM 190090) and JAK (see MIM 147795) families, mediate intracellular signaling downstream of receptor activation.[supplied by OMIM]
Sequence: WPERKPPHNHPMGMPGARKAAALSGGLLSDPELQRKIMEMELSVHWVTHQECQTALGATGGDVASAIRNLKVDQLFLLSSRSRADCWRILEHYQWDLSAASRYVLARPSpecifications
NP_003976 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC46193 | |
TNK1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8711 | |
TNK1 (Human) Recombinant Protein (Q01) | |
WPERKPPHNHPMGMPGARKAAALSGGLLSDPELQRKIMEMELSVHWVTHQECQTALGATGGDVASAIRNLKVDQLFLLSSRSRADCWRILEHYQWDLSAASRYVLARP | |
RUO | |
TNK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |