missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TLE4 Full-length ORF (NP_008936.2, 1 a.a. - 773 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007091-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101432

  • 335.00€ / 10µg
Estimated Shipment: 04-09-2024
to see stock.
Add to basket

Description

Description

Sequence: MIRDLSKMYPQTRHPAPHQPAQPFKFTISESCDRIKEEFQFLQAQYHSLKLECEKLASEKTEMQRHYVMYYEMSYGLNIEMHKQAEIVKRLNAICAQVIPFLSQEHQQQVVQAVERAKQVTMAELNAIIGQQLQAQHLSHGHGLPVPLTPHPSGLQPPAIPPIGSSAGLLALSSALGGQSHLPIKDEKKHHDNDHQRDRDSIKSSSVSPSASFRGAEKHRNSADYSSESKKQKTEEKEIAARYDSDGEKSDDNLVVDVSNEDPSSPRGSPAHSPRENGLDKTRLLKKDAPISPASIASSSSTPSSKSKELSLNEKSTTPVSKSNTPTPRTDAPTPGSNSTPGLRPVPGKPPGVDPLASSLRTPMAVPCPYPTPFGIVPHAGMNGELTSPGAAYAGLHNISPQMSAAAAAAAAAAAYGRSPVVGFDPHHHMRVPAIPPNLTGIPGGKPAYSFHVSADGQMQPVPFPPDALIGPGIPRHARQINTLNHGEVVCAVTISNPTRHVYTGGKGCVKVWDISHPGNKSPVSQLDCLNRDNYIRSCRLLPDGRTLIVGGEASTLSIWDLAAPTPRIKAELTSSAPACYALAISPDSKVCFSCCSDGNIAVWDLHNQTLVRQFQGHTDGASCIDISNDGTKLWTGGLDNTVRSWDLREGRQLQQHDFTSQIFSLGYCPTGEWLAVGMENSNVEVLHVTKPDKYQLHLHESCVLSLKFAHCGKWFVSTGKDNLLNAWRTPYGASIFQSKESSSVLSCDISVDDKYIVTGSGDKKATVYEVIY
Specifications

Specifications

NP_008936.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
110.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCE-1/BCE1/E(spI)/ESG/ESG4/GRG4
TLE4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7091
TLE4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIRDLSKMYPQTRHPAPHQPAQPFKFTISESCDRIKEEFQFLQAQYHSLKLECEKLASEKTEMQRHYVMYYEMSYGLNIEMHKQAEIVKRLNAICAQVIPFLSQEHQQQVVQAVERAKQVTMAELNAIIGQQLQAQHLSHGHGLPVPLTPHPSGLQPPAIPPIGSSAGLLALSSALGGQSHLPIKDEKKHHDNDHQRDRDSIKSSSVSPSASFRGAEKHRNSADYSSESKKQKTEEKEIAARYDSDGEKSDDNLVVDVSNEDPSSPRGSPAHSPRENGLDKTRLLKKDAPISPASIASSSSTPSSKSKELSLNEKSTTPVSKSNTPTPRTDAPTPGSNSTPGLRPVPGKPPGVDPLASSLRTPMAVPCPYPTPFGIVPHAGMNGELTSPGAAYAGLHNISPQMSAAAAAAAAAAAYGRSPVVGFDPHHHMRVPAIPPNLTGIPGGKPAYSFHVSADGQMQPVPFPPDALIGPGIPRHARQINTLNHGEVVCAVTISNPTRHVYTGGKGCVKVWDISHPGNKSPVSQLDCLNRDNYIRSCRLLPDGRTLIVGGEASTLSIWDLAAPTPRIKAELTSSAPACYALAISPDSKVCFSCCSDGNIAVWDLHNQTLVRQFQGHTDGASCIDISNDGTKLWTGGLDNTVRSWDLREGRQLQQHDFTSQIFSLGYCPTGEWLAVGMENSNVEVLHVTKPDKYQLHLHESCVLSLKFAHCGKWFVSTGKDNLLNAWRTPYGASIFQSKESSSVLSCDISVDDKYIVTGSGDKKATVYEVIY
RUO
TLE4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers