missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Human Thymulin Synthetic Peptide, Blocking Peptide, >95% Purity, 100 μg
Description
- Protein: Thymulin (Serum thymic factor)
- Type: Synthetic peptide
- Purity: >95%
- Tested Applications: BL
- Species Reactivity: Human
- Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Blocker and Diluent |
| Content And Storage | Store at -80°C. |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Thymulin |
| Cross Reactivity | Human |
| Quantity | 100 μg |
| For Use With (Application) | Blocking |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?