Learn More
Human Thrombopoietin/Tpo Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11609-SP
This item is not returnable.
View return policy
Description
Thrombopoietin/THPO Monoclonal antibody specifically detects Thrombopoietin/THPO in Human samples. It is validated for Western Blot
Specifications
| Thrombopoietin/THPO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Thrombopoietin/Tpo, Ser22-Gly353, Accession # P40225 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 1065103 | |
| Western Blot 2 μg/mL | |
| P40225 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7066 | |
| Human | |
| Purified |