missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TGIF2LX Partial ORF (NP_620410, 101 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_620410 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 90316 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101107
|
Abnova™
H00090316-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16111107
|
Abnova™
H00090316-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. A homolog of this gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition. [provided by RefSeq]
Sequence: FINARRRILPDMLQQRRNDPIIGHKTGKDAHATHLQSTEASVPAKSGPSGPDNVQSLPLWPLPKGQMSREKQPDPESAPSQKLTGIAQPKSpecifications
NP_620410 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC34726/TGIFLX | |
TGIF2LX | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
90316 | |
TGIF2LX (Human) Recombinant Protein (Q01) | |
FINARRRILPDMLQQRRNDPIIGHKTGKDAHATHLQSTEASVPAKSGPSGPDNVQSLPLWPLPKGQMSREKQPDPESAPSQKLTGIAQPK | |
RUO | |
TGIF2LX | |
Wheat Germ (in vitro) | |
GST | |
Liquid |