Learn More
Abnova™ Human TGIF2LX Partial ORF (NP_620410, 101 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00090316-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. A homolog of this gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition. [provided by RefSeq]
Sequence: FINARRRILPDMLQQRRNDPIIGHKTGKDAHATHLQSTEASVPAKSGPSGPDNVQSLPLWPLPKGQMSREKQPDPESAPSQKLTGIAQPKSpecifications
NP_620410 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FINARRRILPDMLQQRRNDPIIGHKTGKDAHATHLQSTEASVPAKSGPSGPDNVQSLPLWPLPKGQMSREKQPDPESAPSQKLTGIAQPK | |
RUO | |
TGIF2LX | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
90316 | |
TGIF2LX (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC34726/TGIFLX | |
TGIF2LX | |
Recombinant | |
wheat germ expression system |