missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TGF-beta 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10502-SP
This item is not returnable.
View return policy
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| TGF-beta 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Chinese Hamster Ovary cell line, CHO-derived human TGF-beta 1 protein, Ala279-Ser390, Accession # P01137 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 1018746 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P01137 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7040 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction