Learn More
Abnova™ Human TBX6 Partial ORF (NP_004599, 191 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006911-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Knockout studies in mice indicate that this gene is important for specification of paraxial mesoderm structures. [provided by RefSeq]
Sequence: SFHRVKLTNSTLDPHGHLILHSMHKYQPRIHLVRAAQLCSQHWGGMASFRFPETTFISVTAYQNPQITQLKIAANPFAKGFRENGRNCKRERDARVKRKLRGPEPAATESpecifications
NP_004599 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SFHRVKLTNSTLDPHGHLILHSMHKYQPRIHLVRAAQLCSQHWGGMASFRFPETTFISVTAYQNPQITQLKIAANPFAKGFRENGRNCKRERDARVKRKLRGPEPAATE | |
RUO | |
TBX6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6911 | |
TBX6 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB67 | |
TBX6 | |
Recombinant | |
wheat germ expression system |