missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TBK1 Antibody, R&D Systems™

Sheep Antibody
193.00€ - 573.00€
Specifications
| Antigen | TBK1 |
|---|---|
| Dilution | Simple Western 20 ug/mL, Immunohistochemistry 3-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719626
![]() |
R&D Systems
AF9934-SP |
25 μg |
193.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706076
![]() |
R&D Systems
AF9934-100 |
100 μg |
573.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBK1 specifically detects TBK1 in Human samples. It is validated for Immunohistochemistry.Specifications
| TBK1 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Sheep | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 2.7.11, EC 2.7.11.1, FLJ11330, FTDALS4, NAK, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, NFKB-Activating Kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1 | |
| E. coli-derived recombinant human TBK-1, Met1-Gly121, Accession No. Q9UHD2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TBK-1 in direct ELISAs. |
| Simple Western 20 ug/mL, Immunohistochemistry 3-15 ug/mL | |
| Polyclonal | |
| Lyophilized | |
| Human | |
| Q9UHD2 | |
| 29110 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title