missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TAF1A Full-length ORF (NP_005672.1, 1 a.a. - 450 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16109735
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16109735 25 μg 25µg
16199725 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16109735 Supplier Abnova™ Supplier No. H00009015P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the smallest SL1-specific TAF. Two transcripts encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MSDFSEELKGPVTDDEEVETSVLSGAGMHFPWLQTYVETVAIGGKRRKDFAQTTSACLSFIQEALLKHQWQQAAEYMYSYFQTLEDSDSYKRQAAPEIIWKLGSEILFYHPKSNMESFNTFANRMKNIGVMNYLKISLQHALYLLHHGMLKDAKRNLSEAETWRHGENTSSREILINLIQAYKGLLQYYTWSEKKMELSKLDKDDYAYNAVAQDVFNHSWKTSANISALIKIPGVWDPFVKSYVEMLEFYGDRDGAQEVLTNYAYDEKFPSNPNAHIYLYNFLKRQKAPRSKLISVLKILYQIVPSHKLMLEFHTLLRKSEKEEHRKLGLEVLFGVLDFAGCTKNITAWKYLAKYLKNILMGNHLAWVQEEWNSRKNWWPGFHFSYFWAKSDWKEDTALACEKAFVAGLLLGKGCRYFRYILKQDHQILGKKIKRMKRSVKKYSIVNPRL

Specifications

Accession Number NP_005672.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9015
Molecular Weight (g/mol) 79.1kDa
Name TAF1A (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSDFSEELKGPVTDDEEVETSVLSGAGMHFPWLQTYVETVAIGGKRRKDFAQTTSACLSFIQEALLKHQWQQAAEYMYSYFQTLEDSDSYKRQAAPEIIWKLGSEILFYHPKSNMESFNTFANRMKNIGVMNYLKISLQHALYLLHHGMLKDAKRNLSEAETWRHGENTSSREILINLIQAYKGLLQYYTWSEKKMELSKLDKDDYAYNAVAQDVFNHSWKTSANISALIKIPGVWDPFVKSYVEMLEFYGDRDGAQEVLTNYAYDEKFPSNPNAHIYLYNFLKRQKAPRSKLISVLKILYQIVPSHKLMLEFHTLLRKSEKEEHRKLGLEVLFGVLDFAGCTKNITAWKYLAKYLKNILMGNHLAWVQEEWNSRKNWWPGFHFSYFWAKSDWKEDTALACEKAFVAGLLLGKGCRYFRYILKQDHQILGKKIKRMKRSVKKYSIVNPRL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC:17061/RAFI48/SL1/TAFI48
Common Name TAF1A
Gene Symbol TAF1A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.