missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACE/ADAM17 Ectodomain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems MAB9301-500
This item is not returnable.
View return policy
Description
TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready, Knockout Validated.
Specifications
| TACE/ADAM17 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells | |
| P78536 | |
| ADAM17 | |
| Insect ovarian cell line T. ni-derived recombinant human TACE/ADAM17 Pro18-Asn671 Accession # P78536 | |
| 500 μg | |
| Primary | |
| Detects the ectodomain of human TACE/ADAM17 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with the ectodomain of recombinant human ADAM8, 9, 15 and recombinant mouse ADAM10 is observed. | |
| Human | |
| Ascites |
| Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF, KnockDown | |
| 111633 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6868 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction