missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SULT2B1 Full-length ORF (NP_814444.1, 1 a.a. - 365 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16171142
Change view
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16171142 25 ug 25µg
16161142 10 ug 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16171142 Supplier Abnova™ Supplier No. H00006820P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene sulfates dehydroepiandrosterone but not 4-nitrophenol, a typical substrate for the phenol and estrogen sulfotransferase subfamilies. Two alternatively spliced variants that encode different isoforms have been described. [provided by RefSeq]

Sequence: MDGPAEPQIPGLWDTYEDDISEISQKLPGEYFRYKGVPFPVGLYSLESISLAENTQDVRDDDIFIITYPKSGTTWMIEIICLILKEGDPSWIRSVPIWERAPWCETIVGAFSLPDQYSPRLMSSHLPIQIFTKAFFSSKAKVIYMGRNPRDVVVSLYHYSKIAGQLKDPGTPDQFLRDFLKGEVQFGSWFDHIKGWLRMKGKDNFLFITYEELQQDLQGSVERICGFLGRPLGKEALGSVVAHSTFSAMKANTMSNYTLLPPSLLDHRRGAFLRKGVCGDWKNHFTVAQSEAFDRAYRKQMRGMPTFPWDEDPEEDGSPDPEPSPEPEPKPSLEPNTSLEREPRPNSSPSPSPGQASETPHPRPS

Specifications

Accession Number NP_814444.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6820
Molecular Weight (g/mol) 67.7kDa
Name SULT2B1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MDGPAEPQIPGLWDTYEDDISEISQKLPGEYFRYKGVPFPVGLYSLESISLAENTQDVRDDDIFIITYPKSGTTWMIEIICLILKEGDPSWIRSVPIWERAPWCETIVGAFSLPDQYSPRLMSSHLPIQIFTKAFFSSKAKVIYMGRNPRDVVVSLYHYSKIAGQLKDPGTPDQFLRDFLKGEVQFGSWFDHIKGWLRMKGKDNFLFITYEELQQDLQGSVERICGFLGRPLGKEALGSVVAHSTFSAMKANTMSNYTLLPPSLLDHRRGAFLRKGVCGDWKNHFTVAQSEAFDRAYRKQMRGMPTFPWDEDPEEDGSPDPEPSPEPEPKPSLEPNTSLEREPRPNSSPSPSPGQASETPHPRPS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HSST2
Common Name SULT2B1
Gene Symbol SULT2B1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.