missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STRC Partial ORF (NP_714544.1, 25 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00161497-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122247

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein that is associated with the hair bundle of the sensory hair cells in the inner ear. The hair bundle is composed of stiff microvilli called stereocilia and is involved with mechanoreception of sound waves. This gene is part of a tandem duplication on chromosome 15; the second copy is a pseudogene. Mutations in this gene cause autosomal recessive non-syndromic deafness. [provided by RefSeq]

Sequence: APTGPHSLDPGLSFLKSLLSTLDQAPQGSLSRSRFFTFLANISSSFEPGRMGEGPVGEPPPLQPPALRLHDFLVTLRGSPDWEP
Specifications

Specifications

NP_714544.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.98kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
APTGPHSLDPGLSFLKSLLSTLDQAPQGSLSRSRFFTFLANISSSFEPGRMGEGPVGEPPPLQPPALRLHDFLVTLRGSPDWEP
RUO
STRC
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
161497
STRC (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DFNB16/MGC156147
STRC
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers