missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STRC Partial ORF (NP_714544.1, 25 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_714544.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 161497 |
Molecular Weight (g/mol) | 34.98kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16122247
|
Abnova™
H00161497-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16112247
|
Abnova™
H00161497-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein that is associated with the hair bundle of the sensory hair cells in the inner ear. The hair bundle is composed of stiff microvilli called stereocilia and is involved with mechanoreception of sound waves. This gene is part of a tandem duplication on chromosome 15; the second copy is a pseudogene. Mutations in this gene cause autosomal recessive non-syndromic deafness. [provided by RefSeq]
Sequence: APTGPHSLDPGLSFLKSLLSTLDQAPQGSLSRSRFFTFLANISSSFEPGRMGEGPVGEPPPLQPPALRLHDFLVTLRGSPDWEPSpecifications
NP_714544.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB16/MGC156147 | |
STRC | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
161497 | |
STRC (Human) Recombinant Protein (Q01) | |
APTGPHSLDPGLSFLKSLLSTLDQAPQGSLSRSRFFTFLANISSSFEPGRMGEGPVGEPPPLQPPALRLHDFLVTLRGSPDWEP | |
RUO | |
STRC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |