missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STRADB Partial ORF (NP_061041, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055437-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16095555

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein that belongs to the serine/threonine protein kinase STE20 subfamily. One of the active site residues in the protein kinase domain of this protein is altered, and it is thus a pseudokinase. This protein is a component of a complex involved in the activation of serine/threonine kinase 11, a master kinase that regulates cell polarity and energy-generating metabolism. This complex regulates the relocation of this kinase from the nucleus to the cytoplasm, and it is essential for G1 cell cycle arrest mediated by this kinase. The protein encoded by this gene can also interact with the X chromosome-linked inhibitor of apoptosis protein, and this interaction enhances the anti-apoptotic activity of this protein via the JNK1 signal transduction pathway. Two pseudogenes, located on chromosomes 1 and 7, have been found for this gene. [provided by RefSeq]

Sequence: MSLLDCFCTSRTQVESLRPEKQSETSIHQYLVDEPTLSWSRPSTRASEVLCSTNVSHYELQVEIGRGFDNLTSVHLARHTPTGTLVTIKITNLEN
Specifications

Specifications

NP_061041
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLLDCFCTSRTQVESLRPEKQSETSIHQYLVDEPTLSWSRPSTRASEVLCSTNVSHYELQVEIGRGFDNLTSVHLARHTPTGTLVTIKITNLEN
RUO
STRADB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55437
STRADB (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ALS2CR2/CALS-21/ILPIP/ILPIPA/MGC102916/PAPK/PRO1038
STRADB
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers