Learn More
Abnova™ Human STK10 Partial ORF (AAH14413, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006793-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the Ste20 family of serine/threonine protein kinases, and is similar to several known polo-like kinase kinases. The protein can associate with and phosphorylate polo-like kinase 1, and overexpression of a kinase-dead version of the protein interferes with normal cell cycle progression. The kinase can also negatively regulate interleukin 2 expression in T-cells via the mitogen activated protein kinase kinase 1 pathway. [provided by RefSeq]
Sequence: MASFLLFSQFAFHLRQSLVYVGGGDRGKREAEMFILLLPCFMPSRVPLPAHGCECTHIRVHTGLGLLAWPLLPRWVGTRLLPEPVPLSMLGGAVGVARAGCWHRRHGKAQSpecifications
AAH14413 | |
Liquid | |
6793 | |
STK10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LOK/PRO2729 | |
STK10 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASFLLFSQFAFHLRQSLVYVGGGDRGKREAEMFILLLPCFMPSRVPLPAHGCECTHIRVHTGLGLLAWPLLPRWVGTRLLPEPVPLSMLGGAVGVARAGCWHRRHGKAQ | |
RUO | |
STK10 | |
Wheat Germ (in vitro) | |
GST |