missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STK10 Partial ORF (AAH14413, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH14413 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6793 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16106055
|
Abnova™
H00006793-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16116055
|
Abnova™
H00006793-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the Ste20 family of serine/threonine protein kinases, and is similar to several known polo-like kinase kinases. The protein can associate with and phosphorylate polo-like kinase 1, and overexpression of a kinase-dead version of the protein interferes with normal cell cycle progression. The kinase can also negatively regulate interleukin 2 expression in T-cells via the mitogen activated protein kinase kinase 1 pathway. [provided by RefSeq]
Sequence: MASFLLFSQFAFHLRQSLVYVGGGDRGKREAEMFILLLPCFMPSRVPLPAHGCECTHIRVHTGLGLLAWPLLPRWVGTRLLPEPVPLSMLGGAVGVARAGCWHRRHGKAQSpecifications
AAH14413 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LOK/PRO2729 | |
STK10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6793 | |
STK10 (Human) Recombinant Protein (Q01) | |
MASFLLFSQFAFHLRQSLVYVGGGDRGKREAEMFILLLPCFMPSRVPLPAHGCECTHIRVHTGLGLLAWPLLPRWVGTRLLPEPVPLSMLGGAVGVARAGCWHRRHGKAQ | |
RUO | |
STK10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |