missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STARD5 Partial ORF (NP_871629, 121 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080765-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100347

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Cholesterol homeostasis is regulated, at least in part, by sterol regulatory element (SRE)-binding proteins (e.g., SREBP1; MIM 184756) and by liver X receptors (e.g., LXRA; MIM 602423). Upon sterol depletion, LXRs are inactive and SREBPs are cleaved, after which they bind promoter SREs and activate genes involved in cholesterol biosynthesis and uptake. Sterol transport is mediated by vesicles or by soluble protein carriers, such as steroidogenic acute regulatory protein (STAR; MIM 600617). STAR is homologous to a family of proteins containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain, including STARD5 (Soccio et al., 2002 [PubMed 12011452]).[supplied by OMIM]

Sequence: LVKRYEDGTISSNATHVEHPLCPPKPGFVRGFNHPCGCFCEPLPGEPTKTNLVTFFHTDLSGYLPQNVVDSFFPRSMTRFYANLQKAVKQFH
Specifications

Specifications

NP_871629
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.86kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LVKRYEDGTISSNATHVEHPLCPPKPGFVRGFNHPCGCFCEPLPGEPTKTNLVTFFHTDLSGYLPQNVVDSFFPRSMTRFYANLQKAVKQFH
RUO
STARD5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
80765
STARD5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC10327
STARD5
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers