Learn More
Abnova™ Human STARD5 Partial ORF (NP_871629, 121 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_871629 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80765 |
Molecular Weight (g/mol) | 35.86kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100347
|
Abnova™
H00080765-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16190337
|
Abnova™
H00080765-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Cholesterol homeostasis is regulated, at least in part, by sterol regulatory element (SRE)-binding proteins (e.g., SREBP1; MIM 184756) and by liver X receptors (e.g., LXRA; MIM 602423). Upon sterol depletion, LXRs are inactive and SREBPs are cleaved, after which they bind promoter SREs and activate genes involved in cholesterol biosynthesis and uptake. Sterol transport is mediated by vesicles or by soluble protein carriers, such as steroidogenic acute regulatory protein (STAR; MIM 600617). STAR is homologous to a family of proteins containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain, including STARD5 (Soccio et al., 2002 [PubMed 12011452]).[supplied by OMIM]
Sequence: LVKRYEDGTISSNATHVEHPLCPPKPGFVRGFNHPCGCFCEPLPGEPTKTNLVTFFHTDLSGYLPQNVVDSFFPRSMTRFYANLQKAVKQFHSpecifications
NP_871629 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.86kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC10327 | |
STARD5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
80765 | |
STARD5 (Human) Recombinant Protein (Q01) | |
LVKRYEDGTISSNATHVEHPLCPPKPGFVRGFNHPCGCFCEPLPGEPTKTNLVTFFHTDLSGYLPQNVVDSFFPRSMTRFYANLQKAVKQFH | |
RUO | |
STARD5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |