Learn More
Abnova™ Human ST7 Full-length ORF (AAH30954.1, 1 a.a. - 547 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007982-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described. [provided by RefSeq]
Sequence: MAEAATGFLEQLKSCIVWSWTYLWTVWFFIVLFLVYILRVPLKINDNLSTVSMFLNTLTPKFYVALTGTSSLISGLILIFEWWYFRKYGTSFIEQVSVSHLRPLLGGVDNNSSNNSDSNRQSVSECKVWRNPLNLFRGAEYNRYTWVTGREPLTYYDMNLSAQDHQTFFTCDSDHLRPADAIMQKAWRERNPQARISAAHEALEINECATAYILLAEEEATTIAEAEKLFKQALKAGDGCYRRSQQLQHHGSQYEAQHRRDTNVLVYIKRRLAMCARRLGRTREAVKMMRDLMKEFPLLSMFNIHENLLEALLELQAYADVQAVLAKYDDISLPKSATICYTAALLKARAVSDKFSPEAASRRGLSTAEMNAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLAHWKRVEGALNLLHCTWEGTFRMIPYPLEKGHLFYPYPICTETADRELLPSFHEVSVYPKKELPFFILFTAGLCSFTAMLALLTHQFPELMGVFAKAFLSTLFAPLNFVMEKVESILPSSLWHQLTRISpecifications
AAH30954.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp762O2113/ETS7q/FAM4A/FAM4A1/HELG/RAY1/SEN4/TSG7 | |
ST7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7982 | |
ST7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEAATGFLEQLKSCIVWSWTYLWTVWFFIVLFLVYILRVPLKINDNLSTVSMFLNTLTPKFYVALTGTSSLISGLILIFEWWYFRKYGTSFIEQVSVSHLRPLLGGVDNNSSNNSDSNRQSVSECKVWRNPLNLFRGAEYNRYTWVTGREPLTYYDMNLSAQDHQTFFTCDSDHLRPADAIMQKAWRERNPQARISAAHEALEINECATAYILLAEEEATTIAEAEKLFKQALKAGDGCYRRSQQLQHHGSQYEAQHRRDTNVLVYIKRRLAMCARRLGRTREAVKMMRDLMKEFPLLSMFNIHENLLEALLELQAYADVQAVLAKYDDISLPKSATICYTAALLKARAVSDKFSPEAASRRGLSTAEMNAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLAHWKRVEGALNLLHCTWEGTFRMIPYPLEKGHLFYPYPICTETADRELLPSFHEVSVYPKKELPFFILFTAGLCSFTAMLALLTHQFPELMGVFAKAFLSTLFAPLNFVMEKVESILPSSLWHQLTRI | |
RUO | |
ST7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |