missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ST7 Full-length ORF (AAH30954.1, 1 a.a. - 547 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH30954.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7982 |
Molecular Weight (g/mol) | 89.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16122232
|
Abnova™
H00007982-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16112232
|
Abnova™
H00007982-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described. [provided by RefSeq]
Sequence: MAEAATGFLEQLKSCIVWSWTYLWTVWFFIVLFLVYILRVPLKINDNLSTVSMFLNTLTPKFYVALTGTSSLISGLILIFEWWYFRKYGTSFIEQVSVSHLRPLLGGVDNNSSNNSDSNRQSVSECKVWRNPLNLFRGAEYNRYTWVTGREPLTYYDMNLSAQDHQTFFTCDSDHLRPADAIMQKAWRERNPQARISAAHEALEINECATAYILLAEEEATTIAEAEKLFKQALKAGDGCYRRSQQLQHHGSQYEAQHRRDTNVLVYIKRRLAMCARRLGRTREAVKMMRDLMKEFPLLSMFNIHENLLEALLELQAYADVQAVLAKYDDISLPKSATICYTAALLKARAVSDKFSPEAASRRGLSTAEMNAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLAHWKRVEGALNLLHCTWEGTFRMIPYPLEKGHLFYPYPICTETADRELLPSFHEVSVYPKKELPFFILFTAGLCSFTAMLALLTHQFPELMGVFAKAFLSTLFAPLNFVMEKVESILPSSLWHQLTRISpecifications
AAH30954.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAEAATGFLEQLKSCIVWSWTYLWTVWFFIVLFLVYILRVPLKINDNLSTVSMFLNTLTPKFYVALTGTSSLISGLILIFEWWYFRKYGTSFIEQVSVSHLRPLLGGVDNNSSNNSDSNRQSVSECKVWRNPLNLFRGAEYNRYTWVTGREPLTYYDMNLSAQDHQTFFTCDSDHLRPADAIMQKAWRERNPQARISAAHEALEINECATAYILLAEEEATTIAEAEKLFKQALKAGDGCYRRSQQLQHHGSQYEAQHRRDTNVLVYIKRRLAMCARRLGRTREAVKMMRDLMKEFPLLSMFNIHENLLEALLELQAYADVQAVLAKYDDISLPKSATICYTAALLKARAVSDKFSPEAASRRGLSTAEMNAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLAHWKRVEGALNLLHCTWEGTFRMIPYPLEKGHLFYPYPICTETADRELLPSFHEVSVYPKKELPFFILFTAGLCSFTAMLALLTHQFPELMGVFAKAFLSTLFAPLNFVMEKVESILPSSLWHQLTRI | |
RUO | |
ST7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7982 | |
ST7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp762O2113/ETS7q/FAM4A/FAM4A1/HELG/RAY1/SEN4/TSG7 | |
ST7 | |
Recombinant | |
wheat germ expression system |