missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST7 Full-length ORF (AAH30954.1, 1 a.a. - 547 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007982-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122232

  • 508.00€ / 25µg
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described. [provided by RefSeq]

Sequence: MAEAATGFLEQLKSCIVWSWTYLWTVWFFIVLFLVYILRVPLKINDNLSTVSMFLNTLTPKFYVALTGTSSLISGLILIFEWWYFRKYGTSFIEQVSVSHLRPLLGGVDNNSSNNSDSNRQSVSECKVWRNPLNLFRGAEYNRYTWVTGREPLTYYDMNLSAQDHQTFFTCDSDHLRPADAIMQKAWRERNPQARISAAHEALEINECATAYILLAEEEATTIAEAEKLFKQALKAGDGCYRRSQQLQHHGSQYEAQHRRDTNVLVYIKRRLAMCARRLGRTREAVKMMRDLMKEFPLLSMFNIHENLLEALLELQAYADVQAVLAKYDDISLPKSATICYTAALLKARAVSDKFSPEAASRRGLSTAEMNAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLAHWKRVEGALNLLHCTWEGTFRMIPYPLEKGHLFYPYPICTETADRELLPSFHEVSVYPKKELPFFILFTAGLCSFTAMLALLTHQFPELMGVFAKAFLSTLFAPLNFVMEKVESILPSSLWHQLTRI
Specifications

Specifications

AAH30954.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp762O2113/ETS7q/FAM4A/FAM4A1/HELG/RAY1/SEN4/TSG7
ST7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7982
ST7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEAATGFLEQLKSCIVWSWTYLWTVWFFIVLFLVYILRVPLKINDNLSTVSMFLNTLTPKFYVALTGTSSLISGLILIFEWWYFRKYGTSFIEQVSVSHLRPLLGGVDNNSSNNSDSNRQSVSECKVWRNPLNLFRGAEYNRYTWVTGREPLTYYDMNLSAQDHQTFFTCDSDHLRPADAIMQKAWRERNPQARISAAHEALEINECATAYILLAEEEATTIAEAEKLFKQALKAGDGCYRRSQQLQHHGSQYEAQHRRDTNVLVYIKRRLAMCARRLGRTREAVKMMRDLMKEFPLLSMFNIHENLLEALLELQAYADVQAVLAKYDDISLPKSATICYTAALLKARAVSDKFSPEAASRRGLSTAEMNAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLAHWKRVEGALNLLHCTWEGTFRMIPYPLEKGHLFYPYPICTETADRELLPSFHEVSVYPKKELPFFILFTAGLCSFTAMLALLTHQFPELMGVFAKAFLSTLFAPLNFVMEKVESILPSSLWHQLTRI
RUO
ST7
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers