Learn More
Abnova™ Human ST6GALNAC4 Partial ORF (NP_055218, 91 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_055218 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27090 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115686
|
Abnova™
H00027090-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16105686
|
Abnova™
H00027090-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein prefers glycoproteins rather than glycolipids as substrates and shows restricted substrate specificity, utilizing only the trisaccharide sequence Neu5Ac-alpha-2,3-Gal-beta-1,3-GalNAc. In addition, it is involved in the synthesis of ganglioside GD1A from GM1B. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GAEIDSAECVFRMNQAPTVGFEADVGQRSTLRVVSHTSVPLLLRNYSHYFQKARDTLYMVWGQGRHMDRVLGGRTYRTLLQLTRMYPGLQVYTFTERMMASpecifications
NP_055218 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SIAT3C/SIAT7D/ST6GALNACIV | |
ST6GALNAC4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
27090 | |
ST6GALNAC4 (Human) Recombinant Protein (Q01) | |
GAEIDSAECVFRMNQAPTVGFEADVGQRSTLRVVSHTSVPLLLRNYSHYFQKARDTLYMVWGQGRHMDRVLGGRTYRTLLQLTRMYPGLQVYTFTERMMA | |
RUO | |
ST6GALNAC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |