missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST6GALNAC4 Partial ORF (NP_055218, 91 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027090-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105686

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein prefers glycoproteins rather than glycolipids as substrates and shows restricted substrate specificity, utilizing only the trisaccharide sequence Neu5Ac-alpha-2,3-Gal-beta-1,3-GalNAc. In addition, it is involved in the synthesis of ganglioside GD1A from GM1B. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: GAEIDSAECVFRMNQAPTVGFEADVGQRSTLRVVSHTSVPLLLRNYSHYFQKARDTLYMVWGQGRHMDRVLGGRTYRTLLQLTRMYPGLQVYTFTERMMA
Specifications

Specifications

NP_055218
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GAEIDSAECVFRMNQAPTVGFEADVGQRSTLRVVSHTSVPLLLRNYSHYFQKARDTLYMVWGQGRHMDRVLGGRTYRTLLQLTRMYPGLQVYTFTERMMA
RUO
ST6GALNAC4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27090
ST6GALNAC4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SIAT3C/SIAT7D/ST6GALNACIV
ST6GALNAC4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers