missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST5 Partial ORF (NP_005409, 1038 a.a. - 1135 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006764-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16115935

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene was identified by its ability to suppress the tumorigenicity of Hela cells in nude mice. The protein encoded by this gene contains a C-terminal region that shares similarity with the Rab 3 family of small GTP binding proteins. This protein preferentially binds to the SH3 domain of c-Abl kinase, and acts as a regulator of MAPK1/ERK2 kinase, which may contribute to its ability to reduce the tumorigenic phenotype in cells. Three alternatively spliced transcript variants of this gene encoding distinct isoforms are identified. [provided by RefSeq]

Sequence: VGHYSLFLTQSEKGERAFQREAFRKSVASKSIRRFLEVFMESQMFAGFIQDRELRKCRAKGLFEQRVEQYLEELPDTEQSGMNKFLRGLGNKMKFLHK
Specifications

Specifications

NP_005409
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VGHYSLFLTQSEKGERAFQREAFRKSVASKSIRRFLEVFMESQMFAGFIQDRELRKCRAKGLFEQRVEQYLEELPDTEQSGMNKFLRGLGNKMKFLHK
RUO
ST5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6764
ST5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DENND2B/HTS1/MGC33090/p126
ST5
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers