Learn More
Abnova™ Human ST5 Partial ORF (NP_005409, 1038 a.a. - 1135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005409 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6764 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16125935
|
Abnova™
H00006764-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16115935
|
Abnova™
H00006764-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene was identified by its ability to suppress the tumorigenicity of Hela cells in nude mice. The protein encoded by this gene contains a C-terminal region that shares similarity with the Rab 3 family of small GTP binding proteins. This protein preferentially binds to the SH3 domain of c-Abl kinase, and acts as a regulator of MAPK1/ERK2 kinase, which may contribute to its ability to reduce the tumorigenic phenotype in cells. Three alternatively spliced transcript variants of this gene encoding distinct isoforms are identified. [provided by RefSeq]
Sequence: VGHYSLFLTQSEKGERAFQREAFRKSVASKSIRRFLEVFMESQMFAGFIQDRELRKCRAKGLFEQRVEQYLEELPDTEQSGMNKFLRGLGNKMKFLHKSpecifications
NP_005409 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DENND2B/HTS1/MGC33090/p126 | |
ST5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6764 | |
ST5 (Human) Recombinant Protein (Q01) | |
VGHYSLFLTQSEKGERAFQREAFRKSVASKSIRRFLEVFMESQMFAGFIQDRELRKCRAKGLFEQRVEQYLEELPDTEQSGMNKFLRGLGNKMKFLHK | |
RUO | |
ST5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |