Learn More
Human SOX9 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 48 publications
Brand:  R&D Systems AF3075
Product Code. 18081092
  • ONLINE EXCLUSIVE PRICE
    649.00€
    613.20€ / 100µg
    Save 35.80€
    5% Off
Description
SOX9 Polyclonal specifically detects SOX9 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX9
Polyclonal
Unconjugated
P48436
SOX9
E. coli-derived recombinant human SOX9 Met1-Lys151 Accession # P48436
100 μg
Primary
Detects human SOX9 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human SOX10 is observed.
Human
IgG
Western Blot, Immunocytochemistry
LYOPH
Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL
campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
Goat
Affinity Purified
RUO
6662
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.