missing translation for 'onlineSavingsMsg'
Learn More

Human SMPD3 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18759648
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18759648 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18759648 Supplier R&D Systems Supplier No. MAB7184SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SMPD3/NSMase2 Monoclonal specifically detects SMPD3/NSMase2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPD3/NSMase2
Applications Western Blot
Classification Monoclonal
Clone 758612
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NY59
Gene Alias EC 3.1.4.12, FLJ22593, MGC138443, Neutral sphingomyelinase 2, Neutral sphingomyelinase II, nSMase2, nSMase-2, sphingomyelin phosphodiesterase 3, sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinaseII)
Gene Symbols SMPD3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human SMPD3 Val2-Ala655 Accession # Q9NY59
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55512
Test Specificity Detects human SMPD3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) SMPD1 or rhPP2A is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.