Learn More
Abnova™ Human SLCO4C1 Full-length ORF (AAI52990.1, 1 a.a. - 724 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00353189-P01.25ug
Additional Details : Weight : 0.00010kg
Description
SLCO4C1 belongs to the organic anion transporter (OATP) family. OATPs are involved in the membrane transport of bile acids, conjugated steroids, thyroid hormone, eicosanoids, peptides, and numerous drugs in many tissues (Mikkaichi et al., 2004 [PubMed 14993604]).[supplied by OMIM]
Sequence: MKSAKGIENLAFVPSSPDILRRLSASPSQIEVSALSSDPQRENSQPQELQKPQEPQKSPEPSLPSAPPNVSEEKLRSLSLSEFEEGSYGWRNFHPQCLQRCNTPGGFLLHYCLLAVTQGIVVNGLVNISISTVEKRYEMKSSLTGLISSSYDISFCLLSLFVSFFGERGHKPRWLAFAAFMIGLGALVFSLPQFFSGEYKLGSLFEDTCVTTRNSTSCTSSTSSLSNYLYVFILGQLLLGAGGTPLYTLGTAFLDDSVPTHKSSLYIGTGYAMSILGPAIGYVLGGQLLTIYIDVAMGESTDVTEDDPRWLGAWWIGFLLSWIFAWSLIIPFSCFPKHLPGTAEIQAGKTSQAHQSNSNADVKFGKSIKDFPAALKNLMKNAVFMCLVLSTSSEALITTGFATFLPKFIENQFGLTSSFAATLGGAVLIPGAALGQILGGFLVSKFRMTCKNTMKFALFTSGVALTLSFVFMYAKCENEPFAGVSESYNGTGELGNLIAPCNANCNCSRSYYYPVCGDGVQYFSPCFAGCSNPVAHRKPKVYYNCSCIERKTEITSTAETFGFEAKAGKCETHCAKLPIFLCIFFIVIIFTFMAGTPITVSILRCVNHRQRSLALGIQFMVLRLLGTIPGPIIFGFTIDSTCILWDINDCGIKGACWIYDNIKMAHMLVAISVTCKVITMFFNGFAIFLYKPPPSATDVSFHKENAVVTNVLAEQDLNKIVKEGSpecifications
AAI52990.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
106.59kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OATP-H/OATP-M1/OATP4C1/OATPX/PRO2176/SLC21A20 | |
SLCO4C1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
353189 | |
SLCO4C1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKSAKGIENLAFVPSSPDILRRLSASPSQIEVSALSSDPQRENSQPQELQKPQEPQKSPEPSLPSAPPNVSEEKLRSLSLSEFEEGSYGWRNFHPQCLQRCNTPGGFLLHYCLLAVTQGIVVNGLVNISISTVEKRYEMKSSLTGLISSSYDISFCLLSLFVSFFGERGHKPRWLAFAAFMIGLGALVFSLPQFFSGEYKLGSLFEDTCVTTRNSTSCTSSTSSLSNYLYVFILGQLLLGAGGTPLYTLGTAFLDDSVPTHKSSLYIGTGYAMSILGPAIGYVLGGQLLTIYIDVAMGESTDVTEDDPRWLGAWWIGFLLSWIFAWSLIIPFSCFPKHLPGTAEIQAGKTSQAHQSNSNADVKFGKSIKDFPAALKNLMKNAVFMCLVLSTSSEALITTGFATFLPKFIENQFGLTSSFAATLGGAVLIPGAALGQILGGFLVSKFRMTCKNTMKFALFTSGVALTLSFVFMYAKCENEPFAGVSESYNGTGELGNLIAPCNANCNCSRSYYYPVCGDGVQYFSPCFAGCSNPVAHRKPKVYYNCSCIERKTEITSTAETFGFEAKAGKCETHCAKLPIFLCIFFIVIIFTFMAGTPITVSILRCVNHRQRSLALGIQFMVLRLLGTIPGPIIFGFTIDSTCILWDINDCGIKGACWIYDNIKMAHMLVAISVTCKVITMFFNGFAIFLYKPPPSATDVSFHKENAVVTNVLAEQDLNKIVKEG | |
RUO | |
SLCO4C1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |