missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLCO4C1 Full-length ORF (AAI52990.1, 1 a.a. - 724 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAI52990.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 353189 |
Molecular Weight (g/mol) | 106.59kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100334
|
Abnova™
H00353189-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16110334
|
Abnova™
H00353189-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
SLCO4C1 belongs to the organic anion transporter (OATP) family. OATPs are involved in the membrane transport of bile acids, conjugated steroids, thyroid hormone, eicosanoids, peptides, and numerous drugs in many tissues (Mikkaichi et al., 2004 [PubMed 14993604]).[supplied by OMIM]
Sequence: MKSAKGIENLAFVPSSPDILRRLSASPSQIEVSALSSDPQRENSQPQELQKPQEPQKSPEPSLPSAPPNVSEEKLRSLSLSEFEEGSYGWRNFHPQCLQRCNTPGGFLLHYCLLAVTQGIVVNGLVNISISTVEKRYEMKSSLTGLISSSYDISFCLLSLFVSFFGERGHKPRWLAFAAFMIGLGALVFSLPQFFSGEYKLGSLFEDTCVTTRNSTSCTSSTSSLSNYLYVFILGQLLLGAGGTPLYTLGTAFLDDSVPTHKSSLYIGTGYAMSILGPAIGYVLGGQLLTIYIDVAMGESTDVTEDDPRWLGAWWIGFLLSWIFAWSLIIPFSCFPKHLPGTAEIQAGKTSQAHQSNSNADVKFGKSIKDFPAALKNLMKNAVFMCLVLSTSSEALITTGFATFLPKFIENQFGLTSSFAATLGGAVLIPGAALGQILGGFLVSKFRMTCKNTMKFALFTSGVALTLSFVFMYAKCENEPFAGVSESYNGTGELGNLIAPCNANCNCSRSYYYPVCGDGVQYFSPCFAGCSNPVAHRKPKVYYNCSCIERKTEITSTAETFGFEAKAGKCETHCAKLPIFLCIFFIVIIFTFMAGTPITVSILRCVNHRQRSLALGIQFMVLRLLGTIPGPIIFGFTIDSTCILWDINDCGIKGACWIYDNIKMAHMLVAISVTCKVITMFFNGFAIFLYKPPPSATDVSFHKENAVVTNVLAEQDLNKIVKEGSpecifications
AAI52990.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
106.59kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKSAKGIENLAFVPSSPDILRRLSASPSQIEVSALSSDPQRENSQPQELQKPQEPQKSPEPSLPSAPPNVSEEKLRSLSLSEFEEGSYGWRNFHPQCLQRCNTPGGFLLHYCLLAVTQGIVVNGLVNISISTVEKRYEMKSSLTGLISSSYDISFCLLSLFVSFFGERGHKPRWLAFAAFMIGLGALVFSLPQFFSGEYKLGSLFEDTCVTTRNSTSCTSSTSSLSNYLYVFILGQLLLGAGGTPLYTLGTAFLDDSVPTHKSSLYIGTGYAMSILGPAIGYVLGGQLLTIYIDVAMGESTDVTEDDPRWLGAWWIGFLLSWIFAWSLIIPFSCFPKHLPGTAEIQAGKTSQAHQSNSNADVKFGKSIKDFPAALKNLMKNAVFMCLVLSTSSEALITTGFATFLPKFIENQFGLTSSFAATLGGAVLIPGAALGQILGGFLVSKFRMTCKNTMKFALFTSGVALTLSFVFMYAKCENEPFAGVSESYNGTGELGNLIAPCNANCNCSRSYYYPVCGDGVQYFSPCFAGCSNPVAHRKPKVYYNCSCIERKTEITSTAETFGFEAKAGKCETHCAKLPIFLCIFFIVIIFTFMAGTPITVSILRCVNHRQRSLALGIQFMVLRLLGTIPGPIIFGFTIDSTCILWDINDCGIKGACWIYDNIKMAHMLVAISVTCKVITMFFNGFAIFLYKPPPSATDVSFHKENAVVTNVLAEQDLNKIVKEG | |
RUO | |
SLCO4C1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
353189 | |
SLCO4C1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OATP-H/OATP-M1/OATP4C1/OATPX/PRO2176/SLC21A20 | |
SLCO4C1 | |
Recombinant | |
wheat germ expression system |