missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLCO4C1 Full-length ORF (AAI52990.1, 1 a.a. - 724 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00353189-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100334

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

SLCO4C1 belongs to the organic anion transporter (OATP) family. OATPs are involved in the membrane transport of bile acids, conjugated steroids, thyroid hormone, eicosanoids, peptides, and numerous drugs in many tissues (Mikkaichi et al., 2004 [PubMed 14993604]).[supplied by OMIM]

Sequence: MKSAKGIENLAFVPSSPDILRRLSASPSQIEVSALSSDPQRENSQPQELQKPQEPQKSPEPSLPSAPPNVSEEKLRSLSLSEFEEGSYGWRNFHPQCLQRCNTPGGFLLHYCLLAVTQGIVVNGLVNISISTVEKRYEMKSSLTGLISSSYDISFCLLSLFVSFFGERGHKPRWLAFAAFMIGLGALVFSLPQFFSGEYKLGSLFEDTCVTTRNSTSCTSSTSSLSNYLYVFILGQLLLGAGGTPLYTLGTAFLDDSVPTHKSSLYIGTGYAMSILGPAIGYVLGGQLLTIYIDVAMGESTDVTEDDPRWLGAWWIGFLLSWIFAWSLIIPFSCFPKHLPGTAEIQAGKTSQAHQSNSNADVKFGKSIKDFPAALKNLMKNAVFMCLVLSTSSEALITTGFATFLPKFIENQFGLTSSFAATLGGAVLIPGAALGQILGGFLVSKFRMTCKNTMKFALFTSGVALTLSFVFMYAKCENEPFAGVSESYNGTGELGNLIAPCNANCNCSRSYYYPVCGDGVQYFSPCFAGCSNPVAHRKPKVYYNCSCIERKTEITSTAETFGFEAKAGKCETHCAKLPIFLCIFFIVIIFTFMAGTPITVSILRCVNHRQRSLALGIQFMVLRLLGTIPGPIIFGFTIDSTCILWDINDCGIKGACWIYDNIKMAHMLVAISVTCKVITMFFNGFAIFLYKPPPSATDVSFHKENAVVTNVLAEQDLNKIVKEG
Specifications

Specifications

AAI52990.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
106.59kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OATP-H/OATP-M1/OATP4C1/OATPX/PRO2176/SLC21A20
SLCO4C1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
353189
SLCO4C1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKSAKGIENLAFVPSSPDILRRLSASPSQIEVSALSSDPQRENSQPQELQKPQEPQKSPEPSLPSAPPNVSEEKLRSLSLSEFEEGSYGWRNFHPQCLQRCNTPGGFLLHYCLLAVTQGIVVNGLVNISISTVEKRYEMKSSLTGLISSSYDISFCLLSLFVSFFGERGHKPRWLAFAAFMIGLGALVFSLPQFFSGEYKLGSLFEDTCVTTRNSTSCTSSTSSLSNYLYVFILGQLLLGAGGTPLYTLGTAFLDDSVPTHKSSLYIGTGYAMSILGPAIGYVLGGQLLTIYIDVAMGESTDVTEDDPRWLGAWWIGFLLSWIFAWSLIIPFSCFPKHLPGTAEIQAGKTSQAHQSNSNADVKFGKSIKDFPAALKNLMKNAVFMCLVLSTSSEALITTGFATFLPKFIENQFGLTSSFAATLGGAVLIPGAALGQILGGFLVSKFRMTCKNTMKFALFTSGVALTLSFVFMYAKCENEPFAGVSESYNGTGELGNLIAPCNANCNCSRSYYYPVCGDGVQYFSPCFAGCSNPVAHRKPKVYYNCSCIERKTEITSTAETFGFEAKAGKCETHCAKLPIFLCIFFIVIIFTFMAGTPITVSILRCVNHRQRSLALGIQFMVLRLLGTIPGPIIFGFTIDSTCILWDINDCGIKGACWIYDNIKMAHMLVAISVTCKVITMFFNGFAIFLYKPPPSATDVSFHKENAVVTNVLAEQDLNKIVKEG
RUO
SLCO4C1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers