missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC2A4 Partial ORF (NP_001033, 467 a.a. - 509 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

349.00€ - 528.00€

Specifications

Accession Number NP_001033
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6517
Molecular Weight (g/mol) 30.47kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16115375
View Documents Promotion Details
Abnova™
H00006517-Q01.25UG
25 ug
528.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
16105375
View Documents Promotion Details
Abnova™
H00006517-Q01.10UG
10 ug
349.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
Description

Description

This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM). [provided by RefSeq]

Sequence: RVPETRGRTFDQISAAFHRTPSLLEQEVKPSTELEYLGPDEND
Specifications

Specifications

NP_001033
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
30.47kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GLUT4
SLC2A4
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
6517
SLC2A4 (Human) Recombinant Protein (Q01)
RVPETRGRTFDQISAAFHRTPSLLEQEVKPSTELEYLGPDEND
RUO
SLC2A4
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications