missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC28A2 Partial ORF (NP_004203.1, 1 a.a. - 79 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009153-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MEKASGRQSIALSTVETGTVNLGLELMEKEVEPEGSKRTDAQGHSLGDGLGPSTYQRRSRWPFSKARSFCKTHARLFKKSpecifications
NP_004203.1 | |
Liquid | |
9153 | |
SLC28A2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNT2/HCNT2/HsT17153/MGC138252/SPNT1 | |
SLC28A2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.43kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEKASGRQSIALSTVETGTVNLGLELMEKEVEPEGSKRTDAQGHSLGDGLGPSTYQRRSRWPFSKARSFCKTHARLFKK | |
RUO | |
SLC28A2 | |
Wheat Germ (in vitro) | |
GST |