Learn More
Abnova™ Human SLC26A6 Full-length ORF (NP_599025.2, 1 a.a. - 758 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00065010-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the solute carrier 26 family, whose members encode anion transporter proteins. This particular family member encodes a protein involved in transporting chloride, oxalate, sulfate and bicarbonate. Several alternatively spliced transcript variants of this gene, encoding distinct isoforms, have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MGLADASGPRDTQALLSATQAMDLRRRDYHMERPLLNQEHLEELGRWGSAPRTHQWRTWLQCSRARAYALLLQHLPVLVWLPRYPVRDWLLGDLLSGLSVAIMQLPQGLAYALLAGLPPVFGLYSSFYPVFIYFLFGTSRHISVGTFAVMSVMVGSVTESLAPQALNDSMINETARDAARVQVASTLSVLVGLFQVGLGLIHFGFVVTYLSEPLVRGYTTAAAVQVFVSQLKYVFGLHLSSHSGPLSLIYTVLEVCWKLPQSKVGTVVTAAVAGVVLVVVKLLNDKLQQQLPMPIPGELLTLIGATGISYGMGLKHRFEVDVVGNIPAGLVPPVAPNTQLFSKLVGSAFTIAVVGFAIAISLGKIFALRHGYRVDSNQELVALGLSNLIGGIFQCFPVSCSMSRSLVQESTGGNSQVAGAISSLFILLIIVKLGELFHDLPKAVLAAIIIVNLKGMLRQLSDMRSLWKANRADLLIWLVTFTATILLNLDLGLVVAVIFSLLLVVVRTQMPHYSVLGQVPDTDIYRDVAEYSEAKEVRGVKVFRSSATVYFANAEFYSDALKQRCGVDVDFLISQKKKLLKKQEQLKLKQLQKEEKLRKQAASPKGASVSINVNTSLEDMRSNNVEDCKMMVSSGDKMEDATANGQEDSKAPDGSTLKALGLPQPDFHSLILDLGALSFVDTVCLKSLKNIFHDFREIEVEVYMAACHSPVVSQLEAGHFFDASITKKHLFASVHDAVTFALQHPRPVPDSPVSVTRLSpecifications
NP_599025.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586E1422 | |
SLC26A6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
65010 | |
SLC26A6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGLADASGPRDTQALLSATQAMDLRRRDYHMERPLLNQEHLEELGRWGSAPRTHQWRTWLQCSRARAYALLLQHLPVLVWLPRYPVRDWLLGDLLSGLSVAIMQLPQGLAYALLAGLPPVFGLYSSFYPVFIYFLFGTSRHISVGTFAVMSVMVGSVTESLAPQALNDSMINETARDAARVQVASTLSVLVGLFQVGLGLIHFGFVVTYLSEPLVRGYTTAAAVQVFVSQLKYVFGLHLSSHSGPLSLIYTVLEVCWKLPQSKVGTVVTAAVAGVVLVVVKLLNDKLQQQLPMPIPGELLTLIGATGISYGMGLKHRFEVDVVGNIPAGLVPPVAPNTQLFSKLVGSAFTIAVVGFAIAISLGKIFALRHGYRVDSNQELVALGLSNLIGGIFQCFPVSCSMSRSLVQESTGGNSQVAGAISSLFILLIIVKLGELFHDLPKAVLAAIIIVNLKGMLRQLSDMRSLWKANRADLLIWLVTFTATILLNLDLGLVVAVIFSLLLVVVRTQMPHYSVLGQVPDTDIYRDVAEYSEAKEVRGVKVFRSSATVYFANAEFYSDALKQRCGVDVDFLISQKKKLLKKQEQLKLKQLQKEEKLRKQAASPKGASVSINVNTSLEDMRSNNVEDCKMMVSSGDKMEDATANGQEDSKAPDGSTLKALGLPQPDFHSLILDLGALSFVDTVCLKSLKNIFHDFREIEVEVYMAACHSPVVSQLEAGHFFDASITKKHLFASVHDAVTFALQHPRPVPDSPVSVTRL | |
RUO | |
SLC26A6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |